Learn More
Roboz Surgical GLOVER BLDG CLAMP CRV 7CM 2CM
SDP
Catalog No. 5000574993
Encompass
This curved atraumatic bulldog clamp is a surgical instrument engineered to provide precise control of blood flow to specific organs. It is frequently utilized in procedures such as coronary surgery where temporary vessel occlusion is required. The design features a tension-adjusting screw, allowing for consistent pressure application during use. This device is intended for laboratory and research applications only and is not for human use.
  • Features a tension adjusting screw for precise force control
  • Designed with a curved configuration for improved surgical access
  • Provides consistent occlusive pressure between 240 and 260 grams
  • Includes a 2 cm jaw length for targeted vessel management
  • Measures 7 cm in total overall length
Catalog No. 50-005-74993 Supplier Roboz Surgical Supplier No. RS7541
This curved atraumatic bulldog clamp is a surgical instrument engineered to provide precise control of blood flow to specific organs. It is frequently utilized in procedures such as coronary surgery where temporary vessel occlusion is required. The design features a tension-adjusting screw, allowing for consistent pressure application during use. This device is intended for laboratory and research applications only and is not for human use.
  • Features a tension adjusting screw for precise force control
  • Designed with a curved configuration for improved surgical access
  • Provides consistent occlusive pressure between 240 and 260 grams
  • Includes a 2 cm jaw length for targeted vessel management
  • Measures 7 cm in total overall length