Learn More
World Precision Instrument Glover Bulldog Clamp,7cm ,str
SDP
Catalog No. 502521276
Encompass
  • Curved, smooth tips
  • 7cm long
  • Tension adjustment
Catalog No. 50-252-1276 Supplier World Precision Instrument Supplier No. 503239
  • Curved, smooth tips
  • 7cm long
  • Tension adjustment