Learn More
Med Vet International Jorvet Wild Animal Handling Gloves, Small, J0872N, 1 Pair
Catalog No. 5000495179
Encompass_Preferred
Jorvet's small-sized cat and wildlife handling gloves are designed for maximum dexterity and feel. This provides a high level of penetration protection through a combination of special materials. The hand features a high-quality leather completely lined with new Spectra, which is 10X stronger than Kevlar. The gauntlet has a double layer of DuPont Kevlar Knit that resists sharp teeth, talons, and claws that cover up past the elbow. These animal handling gloves are perfect for handling animals that can not inflict crushing-type bites, such as cats, birds, and small animals. These gloves are not bite-proof. Please use caution.
Catalog No. 50-004-95179 Supplier Med Vet International Supplier No. JOR872
Jorvet's small-sized cat and wildlife handling gloves are designed for maximum dexterity and feel. This provides a high level of penetration protection through a combination of special materials. The hand features a high-quality leather completely lined with new Spectra, which is 10X stronger than Kevlar. The gauntlet has a double layer of DuPont Kevlar Knit that resists sharp teeth, talons, and claws that cover up past the elbow. These animal handling gloves are perfect for handling animals that can not inflict crushing-type bites, such as cats, birds, and small animals. These gloves are not bite-proof. Please use caution.