Learn More
Electron Microscopy Sciences Cryo-Grip™ Waterproof Shoulder Length Gloves, Small
SDP
Catalog No. 501929658
Encompass_Preferred
Made from state-of-the-art materials, these waterproof cryogenic gloves are designed to keep you dry and to wick moisture away from your hands, so you can work in comfort for extended periods of time.
The multi-layered construction allows for a maximum level of grip, dexterity, flexibility, and thermal protection – essential features when function is important and safety is critical.
They offer improved grip on small objects while protecting your hands and arms when working in hazardous, ultra-cold environments.
They are designed to provide a high level of thermal protection where the exposure to cryogenic fluids might exist. Feature a stay-dry lining. Waterproof and breathable. Shoulder length gloves small size.
This item is not returnable. View return policy
Catalog No. 50-192-9658 Supplier Electron Microscopy Sciences Supplier No. 71129S
Made from state-of-the-art materials, these waterproof cryogenic gloves are designed to keep you dry and to wick moisture away from your hands, so you can work in comfort for extended periods of time.
The multi-layered construction allows for a maximum level of grip, dexterity, flexibility, and thermal protection – essential features when function is important and safety is critical.
They offer improved grip on small objects while protecting your hands and arms when working in hazardous, ultra-cold environments.
They are designed to provide a high level of thermal protection where the exposure to cryogenic fluids might exist. Feature a stay-dry lining. Waterproof and breathable. Shoulder length gloves small size.