Learn More
Electron Microscopy Sciences Latex Gloves Powder Free, Large
SDP
Supplier:  Electron Microscopy Sciences 7111620
Encompass_Preferred
Made from 100% natural latex with the addition of Aloe Vera and Vitamin E, offering the softness, tactile sensitivity and skin protection. Economical ambidextrous style, beaded cuff, and are chemically resistant to acids, alcohols, ketones and alkalis. Packed in a convenient dispenser box. Non-sterile. Non-sterile, powder free.
Size: Large
Catalog No. 50-189-9500