Learn More
Electron Microscopy Sciences Latex Gloves Powder Free, Small
SDP
Catalog No. 501899492
Encompass_Preferred
Made from 100% natural latex with the addition of Aloe Vera and Vitamin E, offering the softness, tactile sensitivity and skin protection. Economical ambidextrous style, beaded cuff, and are chemically resistant to acids, alcohols, ketones and alkalis. Packed in a convenient dispenser box. Non-sterile. Non-sterile, powder free.
Size: Small
This item is not returnable. View return policy
Catalog No. 50-189-9492 Supplier Electron Microscopy Sciences Supplier No. 7111420
Made from 100% natural latex with the addition of Aloe Vera and Vitamin E, offering the softness, tactile sensitivity and skin protection. Economical ambidextrous style, beaded cuff, and are chemically resistant to acids, alcohols, ketones and alkalis. Packed in a convenient dispenser box. Non-sterile. Non-sterile, powder free.
Size: Small