Learn More
Genesee Scientific Corporation Genesee N-GEN Nitrile Examination Gloves, Medical-Grade, Powder-Free, 1000 gloves
SDP
Catalog No. NC0265829
Encompass
The Genesee Scientific N-GEN Nitrile Examination Gloves are medium-sized, powder-free, medical-grade gloves designed for various scientific and laboratory tasks. Featuring a soft nitrile formulation for comfort and a 5 mil thickness for durability, they provide excellent tactile sensitivity. The ambidextrous design with textured fingertips ensures optimal grip in wet and dry conditions, making them a reliable choice for users who need a safe barrier without the risk of latex allergies.
This item is not returnable. View return policy
Catalog No. NC0265829 Supplier Genesee Scientific Corporation Supplier No. 44101M
The Genesee Scientific N-GEN Nitrile Examination Gloves are medium-sized, powder-free, medical-grade gloves designed for various scientific and laboratory tasks. Featuring a soft nitrile formulation for comfort and a 5 mil thickness for durability, they provide excellent tactile sensitivity. The ambidextrous design with textured fingertips ensures optimal grip in wet and dry conditions, making them a reliable choice for users who need a safe barrier without the risk of latex allergies.