Learn More
Essendant GLOVESKLNGRDLTX12PRGY
Supplier:  Essendant ANS97432CT
Encompass_Preferred
Ambidextrous with tapered coating on the fingertips for added dexterity Made of ultra-high molecular weight polyethylene (UHMWPE) fiber which enhances breathability and comfort Dotted palms all you to keep a better grip ANSI Cut Resistance- A3 Anti-Vibration- No EN 388 Badge- 4340 Size Type- Large
Catalog No. 50-000-20668