Learn More
MSC Gloveworks Industrial Orange Nitrile Gloves, 8 mil, Diamond Textured, 100/Pk.
Catalog No. 5000792543
Encompass
Gloveworks Industrial Orange Nitrile Gloves are durable, 8-mil thick gloves designed for superior puncture and chemical resistance. Featuring a Raised Diamond Texture (RDT), these powder-free gloves provide enhanced grip in both wet and dry conditions, making them suitable for a wide range of industrial applications.
  • Raised Diamond Texture (RDT) enhances grip and reduces sweating.
  • Nitrile construction provides 8-mil thickness for superior puncture and chemical resistance.
  • High-visibility orange color for safety.
  • Touch screen compatible.
  • Latex-free and powder-free design.
  • Offers superb dexterity, tactile sensitivity, and comfort.
  • Dependable barrier protection for industrial use.
  • Ambidextrous and FDA approved (21 CFR 177.2600).
Catalog No. 50-007-92543 Supplier MSC Supplier No. 98709827
Gloveworks Industrial Orange Nitrile Gloves are durable, 8-mil thick gloves designed for superior puncture and chemical resistance. Featuring a Raised Diamond Texture (RDT), these powder-free gloves provide enhanced grip in both wet and dry conditions, making them suitable for a wide range of industrial applications.
  • Raised Diamond Texture (RDT) enhances grip and reduces sweating.
  • Nitrile construction provides 8-mil thickness for superior puncture and chemical resistance.
  • High-visibility orange color for safety.
  • Touch screen compatible.
  • Latex-free and powder-free design.
  • Offers superb dexterity, tactile sensitivity, and comfort.
  • Dependable barrier protection for industrial use.
  • Ambidextrous and FDA approved (21 CFR 177.2600).