Learn More
Electron Microscopy Sciences Safeknit™ Cut-Resistant Gloves Heavy Duty, Size 10
SDP
Catalog No. 501899449
Encompass_Preferred
High performance cut resistant Safeknit glove is a blend of abrasion-resistant Spectra fiber and other synthetics. The mix of these fibers form a rugged cut resistant barrier that offers the histologist sensitivity for manipulation but with extreme comfort and productivity.
NOTE: These gloves are neither puncture proof nor water resistant. They should not be used to protect against intentional cutting. Avoid use near moving blades. Two strand seamless glove.
Size: 10
Catalog No. 50-189-9449 Supplier Electron Microscopy Sciences Supplier No. 7106510
High performance cut resistant Safeknit glove is a blend of abrasion-resistant Spectra fiber and other synthetics. The mix of these fibers form a rugged cut resistant barrier that offers the histologist sensitivity for manipulation but with extreme comfort and productivity.
NOTE: These gloves are neither puncture proof nor water resistant. They should not be used to protect against intentional cutting. Avoid use near moving blades. Two strand seamless glove.
Size: 10