Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Harris Industries Glow Brite Glow-in-the-Dark Safety-Grade Tape

Catalog No. 19019992
Encompass
Encompass_Preferred
Change view
Click to view available options
Product Type:
Glow-in-the-Dark Tape
Size (English):
2 in. x 10 yd.
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Size (English) Quantity
19-019992 Glow-in-the-Dark Tape 2 in. x 10 yd. 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-019992 Supplier Harris Industries Supplier No. GL002
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Description Safety
Color Ivory
Format Tape
Product Type Glow-in-the-Dark Tape
For Use With (Application) Switches handrails building exits, hazardous areas uneven surfaces, low overheads, emergency equipment, stairways/doorways, fire extinguishers
Quantity 1 Ea.
Size (English) 2 in. x 10 yd.
Length (English) 10 yd.
Width (English) 2 in.
Height (English) 10 yd.
Height (Metric) 9.1 m
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.