Learn More
Bioworld Glow Cyanoblue Dye, 5 mL
Catalog No. 50196777
Encompass_Preferred
Premixed and ready-to-load gel dyes. Glow Loading Dyes generate intense DNA/RNA bands with little background or band distortion and can be used with any type of agarose or acrylamide gels. Glow CyanoBlue dye contains bromophenol blue and xylene cyanol in a ficoll-based solution with ethidium bromide (6X concentration).
Catalog No. 50-196-777 Supplier Bioworld Supplier No. 105700081
Premixed and ready-to-load gel dyes. Glow Loading Dyes generate intense DNA/RNA bands with little background or band distortion and can be used with any type of agarose or acrylamide gels. Glow CyanoBlue dye contains bromophenol blue and xylene cyanol in a ficoll-based solution with ethidium bromide (6X concentration).