Learn More
Research Products International Corp 2X Glow RNA Dye, 1.5ml Volume
SDP
Catalog No. 50212591
Encompass
Glow Loading Dyes provide intense DNA/RNA bands with low background and band distortion. Ideal for use with all agarose or acrylamide gels. Dyes reduce exposure problems associated with ethidium bromide disposal. Glow Dyes are Ficoll based and supplied at 6X concentration in 1.5 ml volume. Glow RNA Dyes are ideal for quick screenings of RNA preps and safer than formaldehyde agarose gels. RNA Loading Dyes are formamide based and available at 2X concentration in 1.5 ml volume.
Catalog No. 50-212-591 Supplier Research Products International Corp Supplier No. 248694
Glow Loading Dyes provide intense DNA/RNA bands with low background and band distortion. Ideal for use with all agarose or acrylamide gels. Dyes reduce exposure problems associated with ethidium bromide disposal. Glow Dyes are Ficoll based and supplied at 6X concentration in 1.5 ml volume. Glow RNA Dyes are ideal for quick screenings of RNA preps and safer than formaldehyde agarose gels. RNA Loading Dyes are formamide based and available at 2X concentration in 1.5 ml volume.