Learn More
Accuform™ Glow-In-The-Dark 3D Projection™ Sign: SEVERE WEATHER SHELTER AREA
Catalog No. 17033232 Shop All AccuformNMC Products
Change view
Click to view available options
Material:
Plastic
Catalog No. Material
17-033-232 Plastic
1 options
Catalog No. 17-033-232 Supplier Accuform™ Supplier No. PSP352
Add to Cart
Add to Cart
Projection™ Signs can be seen from various angles for extra visibility when you need provide identification, location, and direction.
  • Features two panels with the message on each panel
  • Durable, high-impact plastic construction and the flanged edges have pre-drilled mounting holes for easy installation
  • Message is printed on photo-luminescent film and bonded to plastic
  • Glow-in-the-dark sign has 10 hour glow rating and photo-luminescent material can be 'energized' by any light source
Specifications
Finish Non-glare
Material Plastic
Quantity 1 Sign
Style Glow-in-the-dark 3D
Adhesive Type Non-adhesive
Mounting Type Corner Holes
Shape Rectangular
Languages English
Displayed Text SEVERE WEATHER SHELTER AREA
Product Type Projection Sign
Color Black, Green, White