Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ GloWear™ 8006 Hi-Vis Neck Shade

Catalog No. 19910173 Shop All Ergodyne Products
Encompass_Preferred
Change view
Click to view available options
Color:
Lime
Orange
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-910-173 Lime
19-910-172 Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-910-173 Supplier Ergodyne™ Supplier No. 29063

missing translation for 'validMainframeMsg'

GloWear™ 8006 Hi-Vis Neck Shade offers protection from the sun and makes you visible to oncoming traffic.

  • ANSI-compliant polyester solid fabric (3.6 oz.)
  • Reflective binding
  • Hook and loop closures
  • Attaches to hard hat suspension headbands

Specifications

Color Lime
For Use With (Application) Road Construction, Utility Crew, Survey Crew
Material Polyester
Product Type Neck Shade
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.