Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ GloWear™ 8911 Class E Two-Tone Pants

Catalog No. 19820724 Shop All Ergodyne Products
Encompass_Preferred
Change view
Click to view available options
Color:
Orange
Yellow
Size:
Small, Medium
Large, X-Large
2X-Large, 3X-Large
3X-Large
4X-Large, 5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19-820-724 Yellow 4X-Large, 5X-Large
19-820-711 Orange Small, Medium
19-820-712 Orange Large, X-Large
19-820-714 Orange 2X-Large, 3X-Large
19-820-715 Orange 4X-Large, 5X-Large
19-820-721 Yellow Small, Medium
19-820-722 Yellow Large, X-Large
19-820-723 Yellow 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 19-820-724 Supplier Ergodyne™ Supplier No. 22969
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

GloWear™ 8911 Class E Two-Tone Pants are constructed of cool mesh with wide leg openings and hook and loop closures.

  • ANSI-compliant polyester mesh (3.3 oz.)
  • 2 in. ANSI-compliant reflective
  • Reflective set on 4.5 in contrasting background
  • Drawstring waist
  • Wide leg openings with hook and loop closures
  • One right leg cargo pocket

Specifications

Size 4X-Large, 5X-Large
Color Yellow
Closure Type Hook and Loop
Waist Style Drawstring
For Use With (Application) Flagging Crew, Road Construction, Utility Crew, Survey Crew, Construction
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.