Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ GloWear™ 8935 Hi-Vis Ranger Hat

Catalog No. 19820726 Shop All Ergodyne Products
Encompass
Change view
Click to view available options
Color:
Lime
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-820-726 -
19-820-725 Lime
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-820-726 Supplier Ergodyne™ Supplier No. 23262
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Class Headwear Hi-Vis Ranger Hat features a wide brim and vented mesh panels.

  • ANSI-compliant 150D oxford polyester
  • Mesh venting panels
  • 1 in. ANSI-compliant reflective
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.