Learn More
Ergodyne™ GloWear™ Class 3 Pullover Hi-Vis Hooded Sweatshirt with Black Front
Shop All Ergodyne ProductsDescription
- Pouch-style pocket for additional hand warmth and storage
- Bottom front and wrist sections of the sweatshirt are black to prevent a dirt from showing
- Contrasting yellow/lime color provides ultimate visibility
- Two inch ANSI-compliant silver reflective tape on the torso, shoulders and sleeves for effective nighttime visibility
- Dawstring hood and elastic knit waistband and cuffs keep the heat in for all-day warmth
- Ideal for jobsites in high-traffic areas like road construction or anywhere that requires high visibility clothing, and for activities like biking, snowmobiling, snow shoeing, ice fishing
Specifications
Specifications
| Size | Medium |
| Material | Polyester Polar Fleece |
| Color | Hi-vis Lime |
| Certifications/Compliance | ANSI/ISEA 107-2015 Type R, Class 3 compliant |
| For Use With (Application) | Flagging Crew, Road Construction, Utility Crew, Survey Crew, Construction, Railway Workers, Warehouse/Distribution |
| Includes | Drawstring hood |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.