Learn More
Neutec Group GlowZone IR Loop Sterilizer
SDP
Catalog No. 14910419 Shop All Neutec Group Products
Change view
Click to view available options
Product Type:
Loop Sterilizer
Catalog No. Product Type
14-910-419 Loop Sterilizer
1 options
Catalog No. 14-910-419 Supplier Neutec Group Supplier No. BTLF4.002.000
Add to Cart
Add to Cart
Flameless high-speed annealing and sterilizing for using IR-HotSpot technology
  • Infrared loop sterilizer used to anneal or sterilize inoculation loops and delicate micro instruments
  • Sterilized at temperatures from 750 to 1000°C after only  5 to 10 sec.
  • Special quartz glass annealing tube
  • Touchless IR-Sensor technology starts each sterilization process automatically
  • Instantly ready to work without any unnecessary warm-up
  • Adjustable sterilization and cool-down timers for two users
  • Cool-Touch housing made of stainless steel and safety glass
Specifications
Product Type Loop Sterilizer