Learn More
CPC Scientific H-Glu-Gly-Thr-Phe-Thr-Ser-Asp-Val-Ser-Ser-Tyr-Leu-Glu-Gly-Gln-Ala-Ala-Lys-Glu-Phe-Ile-Ala-Trp-Leu-Val-Lys-Gly-Arg-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific GLUC017B
Encompass
GLP-1 (9-36) is a truncated product from the degradation of GLP-1 (7-36) by dipeptidyl peptidase IV (DPP-4). The peptide is an antagonist to the GLP-1 (human) receptor and inhibits hepatic glucose production.ONE-LETTER SEQUENCE: EGTFTSDVSSYLEGQAAKEFIAWLVKGR-NH2MOLECULAR FORMULA: C140H214N36O43MOLECULAR WEIGHT:3089.44STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [ 161748-29-4 ], RESEARCH AREA: DiabetesREFERENCES: Ban, K. et. al. Endocrinol 151, 1520 (2010); John, H. et al. Eur J Med Res 13, 73 (2008); Elahi, D. et. al. Obesity (Silver Spring) 16, 1501 (2008)
Catalog No. 50-210-9389