Learn More
Medchemexpress LLC Glp-1(32-36)amide | 1417302-71-6 | 98.4% | 570.73 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP310210MG
Encompass_Preferred
GLP-1(32-36)amide is a pentapeptide derived from the C-terminus of the glucoregulatory hormone GLP-1. It has been shown to inhibit weight gain and modulate whole-body glucose metabolism in diabetic mice. In vitro studies indicate that it retains cell viability and decreases apoptosis in INS-1 cells. In vivo, it protects islets from damage, inhibits weight gain, and relieves polydipsia symptoms in diabetic mice.
  • Maintains cell viability and reduces apoptosis in INS-1 cells
  • Protects islets from damage
  • Inhibits weight gain
  • Alleviates polydipsia symptoms in diabetic mice
  • Slightly lowers mean glucose levels after a glucose challenge in normal mice
  • Prevents the development of diet-induced obesity and hepatic steatosis in high fat-fed mice
Catalog No. 50-003-96574