Learn More
Bachem GLP-1 (7-36) amide (human, bovine, guinea pig, mouse, rat) Trifluoroacetate
SDP
Supplier:  Bachem 4030663.0500
Encompass_Preferred
GLP-1 (7-36) amide (human, bovine, guinea pig, mouse, rat) Trifluoroacetate, H-His-Ala-Glu-Gly-Thr-Phe-Thr-Ser-Asp-Val-Ser-Ser-Tyr-Leu-Glu-Gly-Gln-Ala-Ala-Lys-Glu-Phe-Ile-Ala-Trp-Leu-Val-Lys-Gly-Arg-NH2, 107444-51-9, C149H226N40O45, Mr 3297.68, 0.5mg. Preproglucagon (98-127) amide (human, bovine, guinea pig, mouse, rat), Glucagon-Like Peptide 1 (7-36) amide (human, bovine, guinea pig, mouse, rat), Proglucagon (78-107) amide (human, bovine, guinea pig, mouse, rat), Glucagon-Like Peptide I (7-36), amide, human. GLP-1 (7-36) amide is secreted from the lower small intestine and shows a strong insulinotropic effect. GLP-1 (7-36) amide is considered as the most important incretin hormone. Its action is mediated by receptors expressed by the endocrine pancreatic B-cells. Considerable interest has focused on the development of this peptide as a therapeutic strategy for non-insulin-dependent (type 2 ) diabetes mellitus and associated neuropathy.
Catalog No. NC9735406