Learn More
Tocris Bioscience™ GLP-1 (7-37)



Description
Specifications
Specifications
| Purity | 0.95 |
| CAS | 106612-94-6 |
| Content And Storage | Store at -20°C |
| Description | Glucagon-Like Peptide 1 Receptor Agonists |
| Molecular Weight (g/mol) | Observed M.W.: 3355.71 |
| Quantity | 1 mg |
| Structural Form | C151H228N40O47 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.