Learn More
Bachem GLP-1 (7-37) (human, bovine, guinea pig, mouse, rat) Trifluoroacetate
SDP
Supplier:  Bachem 4034865.0001
Encompass_Preferred
GLP-1 (7-37) (human, bovine, guinea pig, mouse, rat) Trifluoroacetate, H-His-Ala-Glu-Gly-Thr-Phe-Thr-Ser-Asp-Val-Ser-Ser-Tyr-Leu-Glu-Gly-Gln-Ala-Ala-Lys-Glu-Phe-Ile-Ala-Trp-Leu-Val-Lys-Gly-Arg-Gly-OH, 106612-94-6, C151H228N40O47, Mr 3355.71, 1mg. Glucagon-Like Peptide 1 (7-37), Insulinotropin trifluoroacetate salt, Preproglucagon (98-128), Proglucagon (78-108).
Catalog No. NC9385371