Learn More
Phoenix Pharmaceuticals Inc GLP-1 (7-37) Bioactive Peptide, Insulin Releasing, 95% Purity, 200 µg
Supplier:  Phoenix Pharmaceuticals Inc 02813
Encompass
GLP-1 (7-37) is a highly purified peptide from Phoenix Pharmaceuticals, Inc., critical for metabolic research. It stimulates insulin release, reduces glucagon secretion, and slows gastric emptying, aiding blood glucose regulation. Its versatility across species makes it essential for studies in diabetes and obesity, differentiating it from other products. With ≥ 95% purity and solubility in water, it offers reliable performance in diverse research applications.
Catalog No. NC9899499