Learn More
IBL America Glucagon-like Peptide-1 (GLP-1) (9-36/37) ELISA
Supplier:  IBL America 27788
Encompass_Preferred
Solid phase sandwich ELISA for the determination of GLP-1 (9-36/37) in human, mouse, and rat EDTA plasma and cell culture supernatant. For research use only, not for use in diagnostic procedures.
Catalog No. 50-212-0374