Learn More
CPC Scientific H-His-Asp-Glu-Phe-Glu-Arg-His-Ala-Glu-Gly-Thr-Phe-Thr-Ser-Asp-Val-Ser-Ser-Tyr-Leu-Glu-Gly-Gln-Ala-Ala-Lys-Glu-Phe-Ile-Ala-Trp-Leu-Val-Lys-Gly-Arg-Gly-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific GLUC008B
Encompass
GLP, synthesized by posttranslational processing of proglucagon in the intestine and pancreas, plays an important role in metabolic homeostasis. It is capable of converting intestinal epithelial cells into insulin-producing cells, thus serving as a new putative therapeutic compound for the treatment of diabetes mellitus. ONE-LETTER SEQUENCE: HDEFERHAEGTFTSDVSSYLEGQAAKEFIAWLVKGRGMOLECULAR FORMULA: C186H275N51O59MOLECULAR WEIGHT:4169.6STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [87805-34-3], RESEARCH AREA: DiabetesREFERENCES: G.I. Bell et al., Nature, 304, 368 (1983)
Catalog No. 50-210-9374