Learn More
CPC Scientific H-His-Ala-Glu-Gly-Thr-Phe-Thr-Ser-Asp-Val-Ser-Ser-Tyr-Leu-Glu-Gly-Gln-Ala-Ala-Lys-Glu-Phe-Ile-Ala-Trp-Leu-Val-Lys-Gly-Arg-Gly-OH 0.5MG
SDP
Catalog No. 502109386
Encompass
GLP-1 (7-37) is a insulinotropic peptide generated from by precursor GLP-1 (1-37) after proteolytic cleavage. It is secreted by the L cells of intestinal mucosa after food ingestion.SEQUENCE: H-His-Ala-Glu-Gly-Thr-Phe-Thr-Ser-Asp-Val-Ser-Ser-Tyr-Leu-Glu-Gly-Gln-Ala-Ala-Lys-Glu-Phe-Ile-Ala-Trp-Leu-Val-Lys-Gly-Arg-Gly-OHONE-LETTER SEQUENCE: HAEGTFTSDVSSYLEGQAAKEFIAWLVKGRGMOLECULAR FORMULA: C151H228N40O47MOLECULAR WEIGHT: 3355.7STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [106612-94-6]SYNONYMS: Preproglucagon (98-128), Insulinotropin acetate salt, Proglucagon (78-108), Glucagon-Like Peptide 1 (7-37), GLP-1 (7-37) (human, bovine, guinea pig, mouse, rat)RESEARCH AREA: DiabetesREFERENCES:A.Wettergren et al., Regul. Peptides, 77, 83 (1998)A.B.Damholt et al., Endocrinology, 140, 4800 (1999)J.M.Conlon, Regul. Peptides, 93 3 (2000)D.J.Drucker, Gut, 50, 428 (2002)D.J.Drucker, Gastroenterology, 122, 531 (2002)SKU(s): GLUC-016A, GLUC-016B
Catalog No. 50-210-9386 Supplier CPC Scientific Supplier No. GLUC016A
Add to Cart
Add to Cart
GLP-1 (7-37) is a insulinotropic peptide generated from by precursor GLP-1 (1-37) after proteolytic cleavage. It is secreted by the L cells of intestinal mucosa after food ingestion.SEQUENCE: H-His-Ala-Glu-Gly-Thr-Phe-Thr-Ser-Asp-Val-Ser-Ser-Tyr-Leu-Glu-Gly-Gln-Ala-Ala-Lys-Glu-Phe-Ile-Ala-Trp-Leu-Val-Lys-Gly-Arg-Gly-OHONE-LETTER SEQUENCE: HAEGTFTSDVSSYLEGQAAKEFIAWLVKGRGMOLECULAR FORMULA: C151H228N40O47MOLECULAR WEIGHT: 3355.7STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [106612-94-6]SYNONYMS: Preproglucagon (98-128), Insulinotropin acetate salt, Proglucagon (78-108), Glucagon-Like Peptide 1 (7-37), GLP-1 (7-37) (human, bovine, guinea pig, mouse, rat)RESEARCH AREA: DiabetesREFERENCES:A.Wettergren et al., Regul. Peptides, 77, 83 (1998)A.B.Damholt et al., Endocrinology, 140, 4800 (1999)J.M.Conlon, Regul. Peptides, 93 3 (2000)D.J.Drucker, Gut, 50, 428 (2002)D.J.Drucker, Gastroenterology, 122, 531 (2002)SKU(s): GLUC-016A, GLUC-016B