Learn More
Enzo Life Sciences GLP-1 (Arg-Gly, non-amidated, CT) monoclonal antibody (03) (1 mg)
SDP
Supplier:  Enzo Life Sciences BPDABS046031
Encompass_Preferred
Clone: 03. Isotype: IgG1κ. Immunogen: Synthetic peptide corresponding to aa 9-37 of GLP-1. Species reactivity: Mammalian. Applications: ELISA, WB. Purity: Protein A/G-affinity purified. Formulation: Liquid. In 0.01M phosphate buffer, pH 7.4, containing 0.5M sodium chloride and 15mM sodium azide.
Catalog No. 50-201-2423