Learn More
Enzo Life Sciences GLP-1 (7-37, 7-36, amide, free NT) monoclonal antibody (4) (1 mg)
SDP
Supplier:  Enzo Life Sciences BPDABS033041
Encompass_Preferred
Clone: 4. Isotype: IgG1κ. Immunogen: Synthetic peptide corresponding to aa 7-17 of GLP-1. Species reactivity: Mammalian. Applications: ELISA, IHC. Purity: Protein A/G-affinity purified. Formulation: Liquid. In 0.01M phosphate buffer, pH 7.4, containing 0.5M sodium chloride and 15mM sodium azide.
Catalog No. 50-201-2406