Learn More
Enzo Life Sciences GLP-1 (9-37, 9-36, amide, free NT) monoclonal antibody (53) (1 mg)
SDP
Supplier:  Enzo Life Sciences BPDABS044531
Encompass_Preferred
Clone: 53. Isotype: IgG1κ. Immunogen: Synthetic peptide corresponding to aa 9-18 of GLP-1. Species reactivity: Mammalian. Applications: ELISA, WB. Purity: Protein A-affinity purified. Formulation: Liquid. In 0.01M phosphate buffer, pH 7.4, containing 0.5M sodium chloride and 15mM sodium azide.
Catalog No. 50-201-2422