Learn More
Medchemexpress LLC HY-P1348 1mg Medchemexpress, GLP-1 moiety from Dulaglutide CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP13481MG
Encompass_Preferred
Medchemexpress, HY-P1348 1mg GLP-1 moiety from Dulaglutide CAS: GLP-1 moiety from Dulaglutide is a 31-amino acid fragment of Dulaglutide which is a glucagon-like peptide 1 receptor (GLP-1) agonist, extracted from patent US 20160369010 A1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4184