Learn More
Enzo Life Sciences Glucagon like peptide 1 polyclonal antibody (100µl)
SDP
Supplier:  Enzo Life Sciences BMLGA11760100
Encompass_Preferred
Immunogen: Synthetic peptide corresponding to aa 1-19 of human GLP-1. Source: Purified from rabbit serum. Species reactivity: Mammalian. Applications: IHC. Formulation: Liquid. In PBS containing 0.09% sodium azide.
Catalog No. 50-201-0896