Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC GLP-1 receptor agonist 11 | 2784590-83-4 | C31H31ClFN3O4 | 25 MG
SDP

Catalog No. 5000374687
Encompass_Preferred

GLP-1 receptor agonist 11 (compound 3) is an agonist of GLP Receptor. It is utilized in the study of diseases such as diabetes and non-alcoholic fatty liver disease. This product is intended for research use only.

  • Agonist of GLP receptor
  • Used in research for diabetes and non-alcoholic fatty liver disease
  • Purity: 98.01%
  • Appearance: solid, white to off-white
  • Stable under recommended storage conditions
  • For research use only

Catalog No. 50-003-74687 Supplier Medchemexpress LLC Supplier No. 25MGHY153815
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GLP-1 receptor agonist 11 (compound 3) is an agonist of GLP Receptor. It is utilized in the study of diseases such as diabetes and non-alcoholic fatty liver disease. This product is intended for research use only.

  • Agonist of GLP receptor
  • Used in research for diabetes and non-alcoholic fatty liver disease
  • Purity: 98.01%
  • Appearance: solid, white to off-white
  • Stable under recommended storage conditions
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.