Learn More
Cayman Chemical GLP-1 (1-32) (bullfrog) (trifluoroacetate salt), Purity: ≥98%, 5 mg
SDP
Catalog No. 502945210
Encompass_Preferred
An endogenous incretin hormone; increases intracellular cAMP levels using HEK293 cells expressing human GLP-1R (EC50 = 0.051 nM); reduces blood glucose levels in an OGTT in fasted mice at 30 mg/kg
Catalog No. 50-294-5210 Supplier Cayman Chemical Supplier No. 407185 MG
An endogenous incretin hormone; increases intracellular cAMP levels using HEK293 cells expressing human GLP-1R (EC50 = 0.051 nM); reduces blood glucose levels in an OGTT in fasted mice at 30 mg/kg