Learn More
Medchemexpress LLC HY-P3102 5mg Medchemexpress, GLP-1(32-36)amide CAS:1417302-71-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP31025MG
Encompass_Preferred
Medchemexpress, HY-P3102 5mg GLP-1(32-36)amide CAS:1417302-71-6 GLP-1(32-36)amide, a pentapeptide, derived from the C terminus of the glucoregulatory hormone GLP-1. GLP-1(32-36)amide could inhibit weight gain and modulate whole body glucose metabolism in diabetic mice. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3093