Learn More
Cayman Chemical GLP-17-36 acIdhumnmou 1mg
SDP
Catalog No. 502944881
Encompass_Preferred
A derivative of GLP-1 (7-36) amide; enhances insulin-induced phosphorylation of Akt in 3T3-L1 adipocytes at 100 nM in the presence of a DPP-4 inhibitor; reduces plasma levels of total cholesterol, triglycerides, free fatty acids, LDL, insulin, and leptin, as well as adipose tissue weight, in a mouse model of high-fat diet-induced obesity at 150 UG/kg; decreases blood glucose levels, food intake, and gastric emptying in non-obese and ob/ob mice at 3 mg/kg
Catalog No. 50-294-4881 Supplier Cayman Chemical Supplier No. 412501 MG
A derivative of GLP-1 (7-36) amide; enhances insulin-induced phosphorylation of Akt in 3T3-L1 adipocytes at 100 nM in the presence of a DPP-4 inhibitor; reduces plasma levels of total cholesterol, triglycerides, free fatty acids, LDL, insulin, and leptin, as well as adipose tissue weight, in a mouse model of high-fat diet-induced obesity at 150 UG/kg; decreases blood glucose levels, food intake, and gastric emptying in non-obese and ob/ob mice at 3 mg/kg