Learn More
BPS Bioscience Inc GLP-1R/CRE Reporter HEK293 Cell Line, 2 vials, 2 x 10^6 cells/vial, Recombinant Cell Line, Firefly Luciferase, cAMP Response Element (CRE)
SDP
Supplier:  BPS Bioscience Inc 78176
Encompass
GLP-1R/CRE Reporter HEK293 Cell Line, 2 vials, 2 x 10^6 cells/vial, Recombinant Cell Line, Firefly Luciferase, cAMP Response Element (CRE)
Catalog No. NC2100098
Add to Cart
Request a Quote