Learn More
Genscript Corporation GLP-1R mFc Chimera Human
Catalog No. 5000057525
Encompass
The GLP1R a GPCR is found on pancreatic beta cells and brain neurons It regulates blood sugar by enhancing insulin secretion In humans it is encoded by the gene GLP1R on chromosome 6 GLP1R is part of the glucagon receptor GPCR family and comprises an extracellular domain that binds the C-terminal helix of GLP-1 and a transmembrane domain that binds the N-terminal region of GLP-1 The TMD domain contains polar residues that regulate biased signaling while the transmembrane helical boundaries and extracellular surface trigger biased agonism
Catalog No. 50-000-57525 Supplier Genscript Corporation Supplier No. Z03808500
The GLP1R a GPCR is found on pancreatic beta cells and brain neurons It regulates blood sugar by enhancing insulin secretion In humans it is encoded by the gene GLP1R on chromosome 6 GLP1R is part of the glucagon receptor GPCR family and comprises an extracellular domain that binds the C-terminal helix of GLP-1 and a transmembrane domain that binds the N-terminal region of GLP-1 The TMD domain contains polar residues that regulate biased signaling while the transmembrane helical boundaries and extracellular surface trigger biased agonism