Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC GLP-1R modulator C16 | 875005-43-9 | 98.7% | C21H26ClFN2O3 | 50 MG
SDP

Catalog No. 5000353649
Encompass_Preferred

GLP-1R modulator C16 is an allosteric modulator enhancing GLP-1 binding to GLP-1R via a transmembrane site (EC50 8.43 ± 3.82 μM).

  • Enhances GLP-1 binding to GLP-1R.
  • Acts via a transmembrane site.
  • High purity of 98.7% confirmed by LCMS.
  • Recommended storage conditions for powder: -20°C for 3 years, 4°C for 2 years.
  • Recommended storage conditions for in solvent: -80°C for 6 months, -20°C for 1 month.

Catalog No. 50-003-53649 Supplier Medchemexpress LLC Supplier No. HY14183950MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GLP-1R modulator C16 is an allosteric modulator enhancing GLP-1 binding to GLP-1R via a transmembrane site (EC50 8.43 ± 3.82 μM).

  • Enhances GLP-1 binding to GLP-1R.
  • Acts via a transmembrane site.
  • High purity of 98.7% confirmed by LCMS.
  • Recommended storage conditions for powder: -20°C for 3 years, 4°C for 2 years.
  • Recommended storage conditions for in solvent: -80°C for 6 months, -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.