Learn More
Medchemexpress LLC GLP-1R modulator C16 | 875005-43-9 | 99.6% | 408.89 g·mol⁻1 | C21H26ClFN2O3 | 5 MG
SDP
Catalog No. 5000221261
Encompass_Preferred
GLP-1R modulator C16 is a small-molecule allosteric modulator that enhances GLP-1 binding to the GLP-1 receptor via a transmembrane site (CAS 875005-43-9). It is characterized in vitro with an EC50 of 8.43 ± 3.82 μM, a molecular weight of 408.89 g·mol⁻1, and high chemical purity suitable for research applications.
  • Allosteric modulator of GLP-1 receptor activity.
  • Characterized potency (EC50 8.43 ± 3.82 μM) for in vitro studies.
  • High chemical purity (99.6%) for reproducible results.
  • Compact quantities suitable for early-stage pharmacology.
  • Useful for studying GLP-1R pharmacology and GPCR signaling.
Catalog No. 50-002-21261 Supplier Medchemexpress LLC Supplier No. HY1418395MG
GLP-1R modulator C16 is a small-molecule allosteric modulator that enhances GLP-1 binding to the GLP-1 receptor via a transmembrane site (CAS 875005-43-9). It is characterized in vitro with an EC50 of 8.43 ± 3.82 μM, a molecular weight of 408.89 g·mol⁻1, and high chemical purity suitable for research applications.
  • Allosteric modulator of GLP-1 receptor activity.
  • Characterized potency (EC50 8.43 ± 3.82 μM) for in vitro studies.
  • High chemical purity (99.6%) for reproducible results.
  • Compact quantities suitable for early-stage pharmacology.
  • Useful for studying GLP-1R pharmacology and GPCR signaling.