Learn More
Medchemexpress LLC GLP-1R modulator C5 | 421578-93-0 | 99.8% | C24H21NO3 | 100 MG
SDP
Catalog No. 5000353251
Encompass_Preferred
GLP-1R modulator C5 is an allosteric modulator that enhances GLP-1 binding to GLP-1R via a transmembrane site (EC50 1.59 ± 0.53 μM). It is intended for research use only.
  • Allosteric modulator
  • Enhances GLP-1 binding to GLP-1R
  • Suitable for research applications
  • Targets glucagon receptor (GCGR)
  • Relevant for inflammation or immune system disease research
  • Relevant for metabolic or endocrine disease research
  • Relevant for digestive system disease research
Catalog No. 50-003-53251 Supplier Medchemexpress LLC Supplier No. HY141840100MG
GLP-1R modulator C5 is an allosteric modulator that enhances GLP-1 binding to GLP-1R via a transmembrane site (EC50 1.59 ± 0.53 μM). It is intended for research use only.
  • Allosteric modulator
  • Enhances GLP-1 binding to GLP-1R
  • Suitable for research applications
  • Targets glucagon receptor (GCGR)
  • Relevant for inflammation or immune system disease research
  • Relevant for metabolic or endocrine disease research
  • Relevant for digestive system disease research