Learn More
Medchemexpress LLC GLP-1R modulator C5 | 421578-93-0 | 99.8% | C24H21NO3 | 10MG
SDP
Catalog No. 5000261296
Encompass_Preferred
GLP-1R modulator C5 is a small-molecule allosteric modulator that enhances GLP-1 binding to the GLP-1 receptor via a transmembrane site. It is intended for research use and is characterized by an EC50 of 1.59 ± 0.53 μM, molecular formula C24H21NO3, and molecular weight 371.43 g·mol⁻1.
  • Allosteric enhancer of GLP-1 binding to GLP-1R.
  • Measured EC50 of 1.59 ± 0.53 μM.
  • Available as solid (5-100 mg) and 10 mM solution in DMSO.
  • High purity (99.81% by LCMS).
  • Supplied with certificate of analysis for analytical verification.
Catalog No. 50-002-61296 Supplier Medchemexpress LLC Supplier No. HY14184010MG
GLP-1R modulator C5 is a small-molecule allosteric modulator that enhances GLP-1 binding to the GLP-1 receptor via a transmembrane site. It is intended for research use and is characterized by an EC50 of 1.59 ± 0.53 μM, molecular formula C24H21NO3, and molecular weight 371.43 g·mol⁻1.
  • Allosteric enhancer of GLP-1 binding to GLP-1R.
  • Measured EC50 of 1.59 ± 0.53 μM.
  • Available as solid (5-100 mg) and 10 mM solution in DMSO.
  • High purity (99.81% by LCMS).
  • Supplied with certificate of analysis for analytical verification.