Learn More
Medchemexpress LLC GLP-1R modulator C5 | 421578-93-0 | 99.81% | C24H21NO3 | 50 MG
SDP
Catalog No. 5000356765
Encompass_Preferred
GLP-1R modulator C5 is an allosteric modulator that enhances GLP-1 binding to GLP-1R through a transmembrane site, with an EC50 of 1.59 ± 0.53 μM. It is intended for research use only.
  • Allosteric modulator
  • Enhances GLP-1 binding to GLP-1R
  • Intended for research use only
Catalog No. 50-003-56765 Supplier Medchemexpress LLC Supplier No. HY14184050MG
GLP-1R modulator C5 is an allosteric modulator that enhances GLP-1 binding to GLP-1R through a transmembrane site, with an EC50 of 1.59 ± 0.53 μM. It is intended for research use only.
  • Allosteric modulator
  • Enhances GLP-1 binding to GLP-1R
  • Intended for research use only