Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC GLP-1R modulator L7-028 | 2648317-95-5 | 99.5% | C24H28N2O3 | 25 MG
SDP

Catalog No. 5000357222
Encompass_Preferred

GLP-1R modulator L7-028 is an allosteric modulator that enhances GLP-1 binding to GLP-1R via a transmembrane site (EC50 11.01 ± 2.73 μM).

  • Enhances GLP-1 binding to GLP-1R
  • Functions as an allosteric modulator
  • Acts via a transmembrane site
  • Molecular weight: 392.49
  • Appearance: solid, white to off-white
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In-solvent storage: -80°C for 6 months, -20°C for 1 month
  • Soluble in DMSO: 100 mg/mL (254.78 mM)

Catalog No. 50-003-57222 Supplier Medchemexpress LLC Supplier No. 25MGHY141842
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GLP-1R modulator L7-028 is an allosteric modulator that enhances GLP-1 binding to GLP-1R via a transmembrane site (EC50 11.01 ± 2.73 μM).

  • Enhances GLP-1 binding to GLP-1R
  • Functions as an allosteric modulator
  • Acts via a transmembrane site
  • Molecular weight: 392.49
  • Appearance: solid, white to off-white
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In-solvent storage: -80°C for 6 months, -20°C for 1 month
  • Soluble in DMSO: 100 mg/mL (254.78 mM)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.