Learn More
Medchemexpress LLC GLP-1R modulator L7-028 | 2648317-95-5 | MFCD34602898 | 99.5% | 392.49 | C24H28N2O3 | 5 MG
SDP
Catalog No. 5000221689
Encompass_Preferred
GLP-1R modulator L7-028 is a small-molecule positive allosteric modulator of the glucagon-like peptide-1 receptor (GLP-1R). It enhances GLP-1 binding at a transmembrane site (reported EC50 11.01 ± 2.73 μM) and is supplied for research use only as a solid with high analytical purity for in vitro studies of GLP-1R signaling.
  • Enhances GLP-1 binding via a transmembrane allosteric site.
  • Reported EC50 11.01 ± 2.73 μM for GLP-1R modulation.
  • High analytical purity suitable for research (≈99.5%).
  • Available in small pack sizes for laboratory use.
  • Molecular formula C24H28N2O3 and molecular weight 392.49.
Catalog No. 50-002-21689 Supplier Medchemexpress LLC Supplier No. HY1418425MG
GLP-1R modulator L7-028 is a small-molecule positive allosteric modulator of the glucagon-like peptide-1 receptor (GLP-1R). It enhances GLP-1 binding at a transmembrane site (reported EC50 11.01 ± 2.73 μM) and is supplied for research use only as a solid with high analytical purity for in vitro studies of GLP-1R signaling.
  • Enhances GLP-1 binding via a transmembrane allosteric site.
  • Reported EC50 11.01 ± 2.73 μM for GLP-1R modulation.
  • High analytical purity suitable for research (≈99.5%).
  • Available in small pack sizes for laboratory use.
  • Molecular formula C24H28N2O3 and molecular weight 392.49.