Learn More
Medchemexpress LLC GLP-2(3-33) | 275801-62-2 | >99.8% | 3557.89 | 5 MG
SDP
Catalog No. 5000422333
Encompass_Preferred
GLP-2(3-33) is a peptide naturally generated by dipeptidylpeptidase IV (DPPIV). It functions as a partial agonist on the GLP-2 receptor with an EC50 of 5.8 nM and exhibits potential competitive antagonistic properties. This peptide has shown a binding IC50 of 41 nM in GLP-2 receptor-binding assays.
  • Naturally generated by dipeptidylpeptidase IV (DPPIV)
  • Acts as a partial agonist on GLP-2 receptor (EC50=5.8 nM)
  • Exhibits potential competitive antagonistic properties
  • Demonstrates binding IC50 of 41 nM in GLP-2 receptor-binding assay
  • Increases dyslipidemia and hepatic lipid accumulation in HFD-fed mice in vivo
Catalog No. 50-004-22333 Supplier Medchemexpress LLC Supplier No. HYP26255MG
GLP-2(3-33) is a peptide naturally generated by dipeptidylpeptidase IV (DPPIV). It functions as a partial agonist on the GLP-2 receptor with an EC50 of 5.8 nM and exhibits potential competitive antagonistic properties. This peptide has shown a binding IC50 of 41 nM in GLP-2 receptor-binding assays.
  • Naturally generated by dipeptidylpeptidase IV (DPPIV)
  • Acts as a partial agonist on GLP-2 receptor (EC50=5.8 nM)
  • Exhibits potential competitive antagonistic properties
  • Demonstrates binding IC50 of 41 nM in GLP-2 receptor-binding assay
  • Increases dyslipidemia and hepatic lipid accumulation in HFD-fed mice in vivo