Learn More
Enzo Life Sciences Glucagon like peptide 2 (human) (100µg). CAS: 99120-49-7
SDP
Supplier:  Enzo Life Sciences BMLGP14390100
Encompass_Preferred
Glucagon-like peptide 2 is a 33- or 34-amino acid peptide located at the C-terminus of, and cleaved from, pro-glucagon by prohormone convertase 3 (PC3). GLP-2 is a potent intestinotropic growth factor, the action of which is transduced by G protein-coupled membrane-bound receptors (GLP2-R) found in the gut. GLP-2Rs are also found in the rodent CNS. The main actions of GLP-2 appear to be crypt cell proliferation, inhibition of enterocyte apoptosis, enhancement of nutrient adsorption and certain reparative effects. Applications: antiserum adsorption control; standard or tracer for immunoassay. Alternative name: GLP 2. Purity: ≥90%. Solubility: Soluble in dilute acetic acid at 1mg/mL. Long Term Storage: -20°C.
Catalog No. 50-201-0908