Learn More
Alpco Ltd Mouse GLP-2 ELISA
SDP
Supplier:  Alpco Ltd 48GP2MSE01
Encompass_Preferred
The Mouse GLP-2 ELISA is designed for the determination of GLP-2 in Plasma and Serum mouse samples.
Analytical sensitivity
0.412 ng/mL
Dynamic range
0.412-100 ng/mL
Research Use Only. Not for Use in Diagnostic Procedures.
Catalog No. 50-922-559