Learn More
Eurofins Discoverx ChemiScreen GLP-2 Receptor Stable Cell Line, 2 vials. (HTS164C)
Catalog No. 502005994
Encompass
The ChemiScreen GLP-2 Receptor Stable Cell Line is an engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. GLP-2) based activation of GLP2R GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses GLP2R alongside endogenous high levels of the promiscuous G protein, Gα15, coupling the receptor to the calcium signaling pathway. Upon activation of GLP2R, the resulting calcium activity can be fluorescently assayed. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, visit www.discoverx.com/targets/gpcr-target-biology.
Catalog No. 50-200-5994 Supplier Eurofins Discoverx Supplier No. HTS164C
Add to Cart
Add to Cart
Request a Quote
The ChemiScreen GLP-2 Receptor Stable Cell Line is an engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. GLP-2) based activation of GLP2R GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses GLP2R alongside endogenous high levels of the promiscuous G protein, Gα15, coupling the receptor to the calcium signaling pathway. Upon activation of GLP2R, the resulting calcium activity can be fluorescently assayed. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, visit www.discoverx.com/targets/gpcr-target-biology.